Gene Transcripts

Gene Symbol
Cdan1
Entrez Gene ID
68968
Gene Name
congenital dyserythropoietic anemia, type I (human)
Gene Aliases
1500015A01Rik, AI448026, AW492297, CDA-I, CDA1, CDAI
Chromosome Location
Chr. 2: 120716154 - 120731517 on Build GRCm38
UniGene ID
Mm.2289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026891.2 NP_081167.2 14 2121
GenBank mRNA AK157271.1 - 14 2122
BC117555.1 - 12 1406 40098039

View Full Product Details