Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027156.3 | NP_081432.2 | 10, 11 | 1522 | |
GenBank mRNA | AK010015.1 | - | 1, 2 | 126 | |
AK141361.1 | - | 10, 11 | 1503 | ||
AK152599.1 | - | 10, 11 | 1490 | ||
AK211681.1 | - | 3, 4 | 347 | ||
BC023700.1 | - | 7, 8 | 785 | ||
BC060646.1 | - | 10, 11 | 1499 | 6853826 | |
BC069876.1 | - | 3, 4 | 350 |