Gene Transcripts

Gene Symbol
Asnsd1
Entrez Gene ID
70396
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
2210409M21Rik
Chromosome Location
Chr. 1: 53344617 - 53352752 on Build GRCm38
UniGene ID
Mm.285783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290984.1 NP_001277913.1 5 479
NM_133728.3 NP_598489.2 5 1910
GenBank mRNA AK031295.1 - 5 1868
AK034746.1 - 1829
AK049194.1 - 5 1857
AK050045.1 - 5 1850
AK076793.1 - 5 1812
AK155030.1 - 5 1849
AK168103.1 - 5 1850
AK168534.1 - 5 480
AK170510.1 - 5 1826
BC013617.1 - 5 1730 3979452

View Full Product Details