Gene Transcripts

Gene Symbol
Stk26
Entrez Gene ID
70415
Gene Name
serine/threonine kinase 26
Gene Aliases
2610018G03Rik, Mst4
Chromosome Location
Chr. X: 50829694 - 50893098 on Build GRCm38
UniGene ID
Mm.391246
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133729.1 NP_598490.1 6 689
GenBank mRNA AK011445.1 - 6 1073
AK029278.1 - 4 1294
AK034609.1 - 6 728
BC005708.1 - 6 689 2598933
BC132438.1 - 5 492 40130715
BC138193.1 - 5 492 8861212
BC145189.1 - 5 492

View Full Product Details