Gene Transcripts

Gene Symbol
Nol8
Entrez Gene ID
70930
Gene Name
nucleolar protein 8
Gene Aliases
4921532D18Rik, 5730412B09Rik
Chromosome Location
Chr. 13: 49653078 - 49679016 on Build GRCm38
UniGene ID
Mm.258915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271397.1 NP_001258326.1 7 1412
GenBank mRNA AK014994.1 - 7 1413
AK029961.1 - 7 1125
AK147172.1 - 7 1148
BC049166.1 - 7 1003
BC058699.1 - 7 1568
BC125575.1 - 7 1138 40130090
BC125579.1 - 7 1138 40130094

View Full Product Details