Gene Transcripts

Gene Symbol
Vps11
Entrez Gene ID
71732
Gene Name
vacuolar protein sorting 11 (yeast)
Gene Aliases
1200011A11Rik
Chromosome Location
Chr. 9: 44347881 - 44361675 on Build GRCm38
UniGene ID
Mm.295013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027889.1 NP_082165.1 7 1205
GenBank mRNA AK004695.1 - 7 1180
AK032484.1 - 7 1170
AK037658.1 - 6 1556
AK047228.1 - 7 1013
AK090030.1 - 7 1186
AK153963.1 - 7 1178
AK162897.1 - 7 1180
BC029004.1 - 7 1205 5360111

View Full Product Details