Gene Transcripts
- Gene Symbol
- Dnajc6
- Entrez Gene ID
- 72685
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 6
- Gene Aliases
- 2810027M23Rik, mKIAA0473
- Chromosome Location
- Chr. 4: 101496022 - 101645004 on Build GRCm38
- UniGene ID
- Mm.76494
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164583.1 |
NP_001158055.1
|
13, 14 |
2104 |
|
NM_001164584.1 |
NP_001158056.1
|
13, 14 |
2049 |
|
NM_001164585.1 |
NP_001158057.1
|
14, 15 |
2114 |
|
NM_198412.2 |
NP_940804.1
|
14, 15 |
2023 |
|
GenBank mRNA |
AK012831.1 |
- |
4, 5 |
769 |
|
AK049935.1 |
- |
1, 2 |
66 |
|
AK122293.1 |
- |
13, 14 |
2025 |
|
AK147570.1 |
- |
14, 15 |
1986 |
|
AK147637.1 |
- |
14, 15 |
1986 |
|
AK163657.1 |
- |
13, 14 |
1923 |
|
BC035262.1 |
- |
8, 9 |
1258 |
|
BC060734.1 |
- |
13, 14 |
2104 |
6409942 |
BC064460.1 |
- |
14, 15 |
1992 |
6821822 |
View Full Product Details