Gene Transcripts

Gene Symbol
Zcchc12
Entrez Gene ID
72693
Gene Name
zinc finger, CCHC domain containing 12
Gene Aliases
2810028A01Rik, AV136720, Sizn1
Chromosome Location
Chr. X: 36195904 - 36199158 on Build GRCm38
UniGene ID
Mm.297698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028325.3 NP_082601.1 5 847
GenBank mRNA AK012833.1 - 5 764
AY466375.1 - 2 613
BC049071.1 - 4 743 5701407
BC055714.1 - 4 714 6415226

View Full Product Details