Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028920.2 | NP_083196.2 | 4 | 1702 | |
GenBank mRNA | AK016575.1 | - | 4 | 1703 | |
AK029715.1 | - | 4 | 1654 | ||
AK030190.1 | - | 4 | 1703 | ||
BC151178.1 | - | 4 | 1640 | 9088067 | |
BC151179.1 | - | 4 | 1640 | 9088068 |