Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029098.3 | NP_083374.1 | 6 | 636 | |
GenBank mRNA | AK003656.1 | - | 6 | 568 | |
AK030589.1 | - | 6 | 602 | ||
AK049110.1 | - | 6 | 637 | ||
AK143891.1 | - | 6 | 597 | ||
AK154968.1 | - | 6 | 770 | ||
AK170328.1 | - | 6 | 597 | ||
AK173121.1 | - | 6 | 1017 | ||
AY052398.1 | - | 6 | 479 | ||
BC023397.1 | - | 6 | 573 | 5043877 |