Gene Transcripts

Gene Symbol
Rprd2
Entrez Gene ID
75137
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
2810036A19Rik, 4930535B03Rik, 6720469I21Rik, AL022841, AL022940, AU021304, BB077382, mKIAA0460
Chromosome Location
Chr. 3: 95759873 - 95819723 on Build GRCm38
UniGene ID
Mm.196275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081293.1 NP_001074762.1 11 3356
GenBank mRNA AK129150.1 - 10 2903
BC067054.1 - 11 3161 30542928

View Full Product Details