Gene Transcripts

Gene Symbol
Gns
Entrez Gene ID
75612
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
2610016K11Rik, AU042285, C87209, G6S, N28088
Chromosome Location
Chr. 10: 121365067 - 121397253 on Build GRCm38
UniGene ID
Mm.207683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029364.3 NP_083640.1 3, 4 529
GenBank mRNA AK030773.1 - 3, 4 528
AK049162.1 - 3, 4 530
AK054046.1 - 3, 4 529
AK083597.1 - 3, 4 507
AK149863.1 - 3, 4 514
AK151037.1 - 1, 2 151
AK159562.1 - 3, 4 496
AK165180.1 - 3, 4 509
AK165917.1 - 3, 4 479
AK169485.1 - 3, 4 511
AK170791.1 - 3, 4 525
BC055328.1 - 3, 4 523 6400020

View Full Product Details