Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030205.4 | NP_084481.3 | 1 | 122 | |
GenBank mRNA | AK018739.1 | - | 1 | 90 | |
AK037894.1 | - | 1 | 116 | ||
AK039585.1 | - | 1 | 179 | ||
AK040408.1 | - | 128 | |||
AK041000.1 | - | 1 | 95 | ||
AK088155.1 | - | 1 | 90 | ||
AK143624.1 | - | 1 | 81 | ||
AK150051.1 | - | 145 | |||
BC061006.1 | - | 1 | 122 | 6536106 |