Gene Transcripts

Gene Symbol
Klf7
Entrez Gene ID
93691
Gene Name
Kruppel-like factor 7 (ubiquitous)
Gene Aliases
9830124P08Rik
Chromosome Location
Chr. 1: 64033727 - 64122341 on Build GRCm38
UniGene ID
Mm.29466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033563.2 NP_291041.2 2 666
GenBank mRNA AB221646.1 - 2 278
AF327568.1 - 2 557
AF338369.1 - 2 701
AK149362.1 - 3 457
BC004700.1 - 3 431 3499191
BC019604.1 - 2 666 3601534
BC040794.1 - 3 431 3499191

View Full Product Details