Gene Transcripts

Gene Symbol
Usp6nl
Entrez Gene ID
98910
Gene Name
USP6 N-terminal like
Gene Aliases
AI316785, RNTRE, TRE2NL, mKIAA0019
Chromosome Location
Chr. 2: 6322757 - 6444363 on Build GRCm38
UniGene ID
Mm.40673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080548.1 NP_001074017.1 14, 15 1365
NM_181399.3 NP_852064.2 13, 14 1204
GenBank mRNA AK084045.1 - 14, 15 1190
AK129037.1 - 14, 15 1217
AK133119.1 - 5, 6 443
AK155073.1 - 14, 15 1366
BC051184.1 - 13, 14 1207
CN455065.1 - 6, 7 594

View Full Product Details