Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145512.4 | NP_663487.1 | 4 | 399 | |
GenBank mRNA | AK036578.1 | - | 4 | 287 | |
AK075812.1 | - | 3 | 229 | ||
AK156973.1 | - | 4 | 290 | ||
AK170590.1 | - | 3 | 204 | ||
BC017549.1 | - | 4 | 278 | 4924681 |