Gene Transcripts

Gene Symbol
Kif5b
Entrez Gene ID
16573
Gene Name
kinesin family member 5B
Gene Aliases
AL022807, Khc, Khcs, Kns1, Ukhc
Chromosome Location
Chr. 18: 6201005 - 6241524 on Build GRCm38
UniGene ID
Mm.223744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008448.3 NP_032474.2 20 2638
GenBank mRNA BC090841.1 - 20 2614
L27153.1 - 20 2232

View Full Product Details