Gene Transcripts
- Gene Symbol
- Mdk
- Entrez Gene ID
- 17242
- Gene Name
- midkine
- Gene Aliases
- MK, Mek
- Chromosome Location
- Chr. 2: 91929805 - 91932297 on Build GRCm38
- UniGene ID
- Mm.906
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012335.2 |
NP_001012335.1
|
3, 4 |
406 |
|
NM_001012336.2 |
NP_001012336.1
|
3, 4 |
403 |
|
NM_001291481.1 |
NP_001278410.1
|
3, 4 |
313 |
|
NM_001291482.1 |
NP_001278411.1
|
2, 3 |
326 |
|
NM_010784.5 |
NP_034914.1
|
3, 4 |
605 |
|
GenBank mRNA |
AK032742.1 |
- |
3, 4 |
309 |
|
AK084457.1 |
- |
3, 4 |
326 |
|
AL364704.1 |
- |
3, 4 |
286 |
|
BC012244.1 |
- |
3, 4 |
274 |
4167496 |
CF728526.1 |
- |
3, 4 |
299 |
|
CF731708.1 |
- |
3, 4 |
402 |
|
CF738400.1 |
- |
3, 4 |
411 |
|
CF740555.1 |
- |
1, 2 |
123 |
|
CJ174056.1 |
- |
3, 4 |
414 |
|
CJ174578.1 |
- |
2, 3 |
201 |
|
M19662.1 |
- |
2, 3 |
484 |
|
M34327.1 |
- |
3, 4 |
279 |
|
M34328.1 |
- |
3, 4 |
590 |
|
M35833.1 |
- |
3, 4 |
279 |
|
View Full Product Details