Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008815.2 | NP_032841.2 | 4, 5 | 462 | |
GenBank mRNA | AK144538.1 | - | 4, 5 | 419 | |
AK144590.1 | - | 4, 5 | 415 | ||
AK144683.1 | - | 2, 3 | 173 | ||
AK158497.1 | - | 3, 4 | 481 | ||
AK166136.1 | - | 4, 5 | 414 | ||
BY731410.1 | - | 4, 5 | 462 | ||
DQ832277.1 | - | 3, 4 | 182 |