Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008815.2 | NP_032841.2 | 13 | 1697 | |
GenBank mRNA | AK144590.1 | - | 13 | 1650 | |
AK144683.1 | - | 11 | 1413 | ||
AK166136.1 | - | 13 | 1633 | ||
BC068248.1 | - | 5 | 2312 | ||
DQ832277.1 | - | 12 | 1402 |