Gene Transcripts
- Gene Symbol
- Ostf1
- Entrez Gene ID
- 20409
- Gene Name
- osteoclast stimulating factor 1
- Gene Aliases
- C78236, SH3P2, Sh3d3
- Chromosome Location
- Chr. 19: 18580364 - 18631813 on Build GRCm38
- UniGene ID
- Mm.172222
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017375.3 |
NP_059071.1
|
3, 4 |
297 |
|
GenBank mRNA |
AK002899.1 |
- |
3, 4 |
276 |
|
AK005525.1 |
- |
3, 4 |
266 |
|
AK008123.1 |
- |
3, 4 |
245 |
|
AK008467.1 |
- |
3, 4 |
245 |
|
AK010074.1 |
- |
3, 4 |
293 |
|
AK145949.1 |
- |
3, 4 |
248 |
|
AK149175.1 |
- |
|
275 |
|
AK150145.1 |
- |
3, 4 |
267 |
|
AK155028.1 |
- |
3, 4 |
267 |
|
BC060986.1 |
- |
3, 4 |
176 |
30300544 |
BY065904.1 |
- |
3, 4 |
297 |
|
U58888.1 |
- |
3, 4 |
245 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC020352.1 |
- |
|
2601 |
|
Chr. - - [-] |
View Full Product Details