Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
210094
Gene Name
IgLON family member 5
Gene Aliases
A230106M20Rik, Gm1431
Chromosome Location
Chr. 7: 43472907 - 43490057 on Build GRCm38
UniGene ID
Mm.228087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164518.1 NP_001157990.1 3 369
GenBank mRNA AK039193.1 - 3 336

View Full Product Details