Gene Transcripts

Gene Symbol
Kcnk12
Entrez Gene ID
210741
Gene Name
potassium channel, subfamily K, member 12
Gene Aliases
mntk1
Chromosome Location
Chr. 17: 87745821 - 87797994 on Build GRCm38
UniGene ID
Mm.312046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199251.1 NP_954859.1 2 1017
GenBank mRNA AB052939.1 - 2 1017
BC138133.1 - 2 809 8861152
BC138134.1 - 2 809 8861153

View Full Product Details