Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252532.1 | NP_001239461.1 | 11 | 1228 | |
NM_145438.2 | NP_663413.2 | 11 | 1228 | ||
GenBank mRNA | AK143486.1 | - | 11 | 1215 | |
AK143798.1 | - | 11 | 1202 | ||
AK167536.1 | - | 11 | 1200 | ||
AY033650.1 | - | 11 | 1178 |