Gene Transcripts
- Gene Symbol
- Eif4e1b
- Entrez Gene ID
- 218268
- Gene Name
- eukaryotic translation initiation factor 4E family member 1B
- Gene Aliases
- Eif4eloo, Gm273
- Chromosome Location
- Chr. 13: 54783927 - 54788459 on Build GRCm38
- UniGene ID
- Mm.425821
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033269.3 |
NP_001028441.1
|
6, 7 |
721 |
|
NM_001039683.2 |
NP_001034772.1
|
6, 7 |
683 |
|
NM_001286178.1 |
NP_001273107.1
|
6, 7 |
650 |
|
NM_001286179.1 |
NP_001273108.1
|
6, 7 |
653 |
|
NM_001286180.1 |
NP_001273109.1
|
6, 7 |
612 |
|
GenBank mRNA |
AK139577.1 |
- |
5, 6 |
959 |
|
BC139143.1 |
- |
6, 7 |
688 |
8862165 |
BC139144.1 |
- |
6, 7 |
685 |
8862166 |
DQ279473.1 |
- |
6, 7 |
716 |
|
DQ279474.1 |
- |
6, 7 |
661 |
|
DQ279478.1 |
- |
6, 7 |
621 |
|
DQ279479.1 |
- |
5, 6 |
554 |
|
DV653822.1 |
- |
6, 7 |
647 |
|
DV659735.1 |
- |
6, 7 |
650 |
|
DV662251.1 |
- |
6, 7 |
650 |
|
DV662389.1 |
- |
6, 7 |
609 |
|
View Full Product Details