Gene Transcripts

Gene Symbol
Oas1e
Entrez Gene ID
231699
Gene Name
2'-5' oligoadenylate synthetase 1E
Gene Aliases
C87484, oasl7
Chromosome Location
Chr. 5: 120786312 - 120795530 on Build GRCm38
UniGene ID
Mm.379383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145210.2 NP_660211.2 2 314
GenBank mRNA AB067531.1 - 2 288
AK087823.1 - 2 314
AY055830.1 - 2 287
AY055831.1 - 2 287
BC053721.1 - 2 311 30053184
BC094918.1 - 2 304 30050168
BC108956.1 - 2 310 40046020

View Full Product Details