Gene Transcripts
- Gene Symbol
- Hsd17b12
- Entrez Gene ID
- 56348
- Gene Name
- hydroxysteroid (17-beta) dehydrogenase 12
- Gene Aliases
- 2610510O05Rik, AI172963, KIK-I, Kik1
- Chromosome Location
- Chr. 2: 94032697 - 94157909 on Build GRCm38
- UniGene ID
- Mm.22505
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019657.4 |
NP_062631.1
|
7, 8 |
569 |
|
GenBank mRNA |
AF064635.1 |
- |
7, 8 |
548 |
|
AK078841.1 |
- |
7, 8 |
570 |
|
AK081869.1 |
- |
7, 8 |
563 |
|
AK082715.1 |
- |
7, 8 |
563 |
|
AK088521.1 |
- |
7, 8 |
563 |
|
AK150849.1 |
- |
7, 8 |
564 |
|
AK152489.1 |
- |
7, 8 |
564 |
|
AK166027.1 |
- |
7, 8 |
563 |
|
AK167908.1 |
- |
7, 8 |
563 |
|
AK188070.1 |
- |
2, 3 |
46 |
|
BC010749.1 |
- |
7, 8 |
544 |
|
BC037620.1 |
- |
6, 7 |
486 |
5375831 |
BC090659.1 |
- |
7, 8 |
539 |
30675153 |
View Full Product Details