Gene Transcripts

Gene Symbol
Mcts2
Entrez Gene ID
66405
Gene Name
malignant T cell amplified sequence 2
Gene Aliases
2400002F11Rik
Chromosome Location
Chr. 2: 152687148 - 152687946 on Build GRCm38
UniGene ID
Mm.440291
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025543.2 NP_079819.1 1 371
GenBank mRNA AK002563.1 - 1 349
AK010255.1 - 1 372
AK028001.1 - 1 350
BC027507.1 - 1 364 1478068

View Full Product Details