Gene Transcripts

Gene Symbol
Trpd52l3
Entrez Gene ID
66745
Gene Name
tumor protein D52-like 3
Gene Aliases
4931412G03Rik
Chromosome Location
Chr. 19: 30003790 - 30006020 on Build GRCm38
UniGene ID
Mm.158128
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025741.2 NP_080017.1 1 263
GenBank mRNA AK016362.1 - 1 252
AK016450.1 - 1 263
BC125314.1 - 1 263 40129829
BC125316.1 - 1 263 40129831

View Full Product Details