Gene Transcripts

Gene Symbol
Efcab1
Entrez Gene ID
66793
Gene Name
EF hand calcium binding domain 1
Gene Aliases
4930449A16Rik, 5430404L10Rik
Chromosome Location
Chr. 16: 14906257 - 14969061 on Build GRCm38
UniGene ID
Mm.100270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025769.3 NP_080045.1 2, 3 331
GenBank mRNA AK015426.1 - 313
AK015866.1 - 2, 3 351
AK017267.1 - 2, 3 332
AK030381.1 - 2, 3 341
AK038735.1 - 2, 3 341
AK042748.1 - 2, 3 373
AK043519.1 - 2, 3 303
AK077722.1 - 2, 3 334
BC085167.1 - 2, 3 223 30538702

View Full Product Details