Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 9 | 1091 | |
GenBank mRNA | AK013129.1 | - | 8 | 920 | |
AK019574.1 | - | 9 | 919 | ||
AK078438.1 | - | 9 | 1072 | ||
AK088705.1 | - | 9 | 1072 | ||
BC020162.1 | - | 885 | 3991990 | ||
BC055848.1 | - | 9 | 2009 |