Gene Transcripts

Gene Symbol
Chmp2a
Entrez Gene ID
68953
Gene Name
charged multivesicular body protein 2A
Gene Aliases
1500016L11Rik
Chromosome Location
Chr. 7: 13032006 - 13034777 on Build GRCm38
UniGene ID
Mm.295670
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026885.3 NP_081161.1 5, 6 728
GenBank mRNA AK005267.1 - 5, 6 696
BC012230.1 - 5, 6 669 4024872

View Full Product Details