Gene Transcripts

Gene Symbol
Ing2
Entrez Gene ID
69260
Gene Name
inhibitor of growth family, member 2
Gene Aliases
2810011M06Rik, Ing1l, Ing2b, P33ING2
Chromosome Location
Chr. 8: 47667178 - 47675609 on Build GRCm38
UniGene ID
Mm.430733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023503.3 NP_075992.2 2 806
GenBank mRNA AB433625.1 - 2 309
AK012716.1 - 2 803
AK048800.1 - 2 802
AK083144.1 - 2 805
BC050003.1 - 2 751
BC096433.1 - 2 786 6310342
BC138229.1 - 2 537 8861248

View Full Product Details