Gene Transcripts

Gene Symbol
Bod1
Entrez Gene ID
69556
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
2310022M17Rik, BOD2.1, Fam44b
Chromosome Location
Chr. 11: 31665150 - 31671862 on Build GRCm38
UniGene ID
Mm.34183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024919.1 NP_001020090.1 2 394
GenBank mRNA AK009485.1 - 2 376
AK219559.1 - 1 45
BB616782.1 - 2 418
BC116435.1 - 3 226 40104481
BC116436.1 - 3 226 40104482
BC128508.1 - 3 296
BC128509.1 - 3 226 40104485

View Full Product Details