Gene Transcripts

Gene Symbol
Ms4a10
Entrez Gene ID
69826
Gene Name
membrane-spanning 4-domains, subfamily A, member 10
Gene Aliases
2010001N17Rik
Chromosome Location
Chr. 19: 10962289 - 10974725 on Build GRCm38
UniGene ID
Mm.273404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023529.2 NP_076018.2 5 482
GenBank mRNA AB026046.1 - 5 491
AF237918.1 - 5 482
AK008019.1 - 3 341
BC144935.1 - 5 479 9053483
BC150675.1 - 5 479 9087586

View Full Product Details