Gene Transcripts

Gene Symbol
Utp23
Entrez Gene ID
78581
Gene Name
UTP23, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
1700010I21Rik, AI662478, BB238373, D530033C11Rik
Chromosome Location
Chr. 15: 51877441 - 51884622 on Build GRCm38
UniGene ID
Mm.176347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030132.5 NP_084408.1 3 700
GenBank mRNA AK005831.1 - 3 501
AK021306.1 - 3 650
AK042518.1 - 3 657
AK134445.1 - 3 632
BC027540.1 - 2 461
BC046791.1 - 3 645 5717427
BC056961.1 - 3 645 5717427

View Full Product Details