Gene Transcripts

Gene Symbol
Btbd19
Entrez Gene ID
78611
Gene Name
BTB (POZ) domain containing 19
Gene Aliases
9530048O09Rik, BB115071
Chromosome Location
Chr. 4: 117119218 - 117125725 on Build GRCm38
UniGene ID
Mm.446227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020599.1 - 2 198
AK141215.1 - 2 285


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_001033187.1 - 285 Chr. - - [-]

View Full Product Details