Gene Transcripts
- Gene Symbol
- Otp
- Entrez Gene ID
- 18420
- Gene Name
- orthopedia homolog (Drosophila)
- Gene Aliases
- Chromosome Location
- Chr. 13: 94875627 - 94885130 on Build GRCm38
- UniGene ID
- Mm.42051
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011021.3 |
NP_035151.1
|
1, 2 |
177 |
|
GenBank mRNA |
Y10413.1 |
- |
1, 2 |
179 |
|
View Full Product Details