Gene Transcripts

Gene Symbol
Asb16
Entrez Gene ID
217217
Gene Name
ankyrin repeat and SOCS box-containing 16
Gene Aliases
Chromosome Location
Chr. 11: 102268823 - 102278061 on Build GRCm38
UniGene ID
Mm.216432
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148953.2 NP_683755.1 1 22
GenBank mRNA AF403044.1 - 1 22
BC127245.1 - 1 18
BC127246.1 - 1 21
BC152348.1 - 1 22
BC152349.1 - 1 22 40104909

View Full Product Details