Gene Transcripts

Gene Symbol
Cytl1
Entrez Gene ID
231162
Gene Name
cytokine-like 1
Gene Aliases
4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147
Chromosome Location
Chr. 5: 37735519 - 37739820 on Build GRCm38
UniGene ID
Mm.76407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081106.1 NP_001074575.1 1 41
GenBank mRNA BC116224.1 - 1 12
BC116225.1 - 1 12 40091369
EF108311.1 - 1 12

View Full Product Details