Gene Transcripts

Gene Symbol
Mmp17
Entrez Gene ID
23948
Gene Name
matrix metallopeptidase 17
Gene Aliases
MT4-MMP, MT4MMP, MTMMP4
Chromosome Location
Chr. 5: 129584174 - 129608211 on Build GRCm38
UniGene ID
Mm.42047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011846.4 NP_035976.3 5 870
GenBank mRNA AB021224.2 - 5 876
AJ010731.1 - 5 874
AK079221.1 - 642
AK143521.1 - 5 869
BC051917.1 - 5 870 5716458

View Full Product Details