Gene Transcripts

Gene Symbol
Tor2a
Entrez Gene ID
30933
Gene Name
torsin family 2, member A
Gene Aliases
Prosalusin
Chromosome Location
Chr. 2: 32757231 - 32762244 on Build GRCm38
UniGene ID
Mm.200695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152800.3 NP_690013.1 3, 4 669
GenBank mRNA AK162719.1 - 3, 4 613
AK170979.1 - 2, 3 1848
AK171247.1 - 3, 4 621
BC003466.1 - 3, 4 654 2649388
BC023085.1 - 1, 2 168
BC024469.1 - 3, 4 632 4976022
CK031473.1 - 3, 4 530

View Full Product Details