Gene Transcripts

Gene Symbol
Prl2a1
Entrez Gene ID
56635
Gene Name
prolactin family 2, subfamily a, member 1
Gene Aliases
PLP-M, Prlpm
Chromosome Location
Chr. 13: 27801655 - 27808716 on Build GRCm38
UniGene ID
Mm.23585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019991.1 NP_064375.1 3 320
GenBank mRNA AB032428.1 - 3 341
AF226610.1 - 3 276
AF234636.1 - 3 320
AF272368.1 - 3 272
AK005424.1 - 3 297
BC099458.1 - 3 316 30790018

View Full Product Details