Gene Transcripts

Gene Symbol
Treml1
Entrez Gene ID
71326
Gene Name
triggering receptor expressed on myeloid cells-like 1
Gene Aliases
5430401J17Rik, TLT-1
Chromosome Location
Chr. 17: 48359916 - 48367176 on Build GRCm38
UniGene ID
Mm.90177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289451.1 NP_001276380.1 5, 6 688
NM_001289457.1 NP_001276386.1 6 791
NM_027763.2 NP_082039.1 6 806
GenBank mRNA AK017256.1 - 6 792
AK079428.1 - 6 790
AY078502.1 - 6 806
AY286071.1 - 6 791
BC138399.1 - 6 790 8861419
BC145263.1 - 6 790 9053816
DQ087181.1 - 6 559
DQ087182.1 - 5 940
DQ087183.1 - 5, 6 685

View Full Product Details