Gene Transcripts

Gene Symbol
Ankrd12
Entrez Gene ID
106585
Gene Name
ankyrin repeat domain 12
Gene Aliases
2900001A12Rik, AI447928, ANCO-2, AV347965, GAC-1, mKIAA0874
Chromosome Location
Chr. 17: 65965703 - 66077046 on Build GRCm38
UniGene ID
Mm.34706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025572.1 NP_001020743.1 9 1529
GenBank mRNA AK134530.1 - 4 708
AK153850.1 - 8 1438
BC023046.1 - 9 1400
BC050185.1 - 9 1509
BC064777.1 - 8 1444
BC080825.1 - 8 1452

View Full Product Details