Gene Transcripts

Gene Symbol
Kctd1
Entrez Gene ID
106931
Gene Name
potassium channel tetramerisation domain containing 1
Gene Aliases
4933402K10Rik, AI661543, AW553000
Chromosome Location
Chr. 18: 14968685 - 15151446 on Build GRCm38
UniGene ID
Mm.329299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142731.1 NP_001136203.1 5 2570
NM_134112.5 NP_598873.2 5 886
GenBank mRNA AK016623.1 - 4 931
AY899808.1 - 5 886
BC022615.1 - 3 554
BC087623.1 - 5 785

View Full Product Details