Gene Transcripts

Gene Symbol
Prdx6
Entrez Gene ID
11758
Gene Name
peroxiredoxin 6
Gene Aliases
1-Cys Prx, 1-cysPrx, 9430088D19Rik, AA690119, Aop2, Brp-12, CP-3, GPx, Ltw-4, Ltw4, Lvtw-4, NSGPx, ORF06, Prdx5, aiPLA2
Chromosome Location
Chr. 1: 161240112 - 161251210 on Build GRCm38
UniGene ID
Mm.186185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF004670.1 - 4 520
AF093852.1 - 4 551
AK030413.1 - 4 553
AK076191.1 - 3 389
AK157820.1 - 4 570
AK161710.1 - 4 553
AK168223.1 - 4 524
AK172888.1 - 4 513
BC013489.1 - 4 512 4215591
BC061181.1 - 4 552 30309446
Y12883.1 - 4 570

View Full Product Details