Gene Transcripts
- Gene Symbol
- Cdk7
- Entrez Gene ID
- 12572
- Gene Name
- cyclin-dependent kinase 7
- Gene Aliases
- AI323415, AI528512, C230069N13, Cdkn7, Crk4
- Chromosome Location
- Chr. 13: 100697024 - 100730942 on Build GRCm38
- UniGene ID
- Mm.259718
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009874.3 |
NP_034004.2
|
7, 8 |
638 |
|
GenBank mRNA |
AK039084.1 |
- |
7, 8 |
626 |
|
AK145759.1 |
- |
7, 8 |
564 |
|
AK168286.1 |
- |
7, 8 |
637 |
|
BC004605.1 |
- |
6, 7 |
485 |
3585145 |
BC068160.1 |
- |
7, 8 |
623 |
30358255 |
BC141043.1 |
- |
7, 8 |
595 |
9055655 |
U11822.1 |
- |
7, 8 |
601 |
|
X57239.1 |
- |
1, 2 |
122 |
|
X74145.1 |
- |
7, 8 |
620 |
|
View Full Product Details