Gene Transcripts
- Gene Symbol
- Klk1b22
- Entrez Gene ID
- 13646
- Gene Name
- kallikrein 1-related peptidase b22
- Gene Aliases
- Egfbp-1, Egfbp1, Klk22, mGk-22
- Chromosome Location
- Chr. 7: 44112683 - 44116876 on Build GRCm38
- UniGene ID
- Mm.5193
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010114.1 |
NP_034244.1
|
2, 3 |
222 |
|
GenBank mRNA |
BC106155.1 |
- |
2, 3 |
226 |
|
View Full Product Details