Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
13830
Gene Name
stomatin
Gene Aliases
Epb7.2
Chromosome Location
Chr. 2: 35313990 - 35337009 on Build GRCm38
UniGene ID
Mm.295284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013515.2 NP_038543.1 2 176
GenBank mRNA AF093620.1 - 2 127
AK077435.1 - 2 144
AK139242.1 - 2 144
AK143724.1 - 2 144
AK148975.1 - 2 145
AK149689.1 - 2 144
AK149821.1 - 2 144
AK149991.1 - 2 144
AK151129.1 - 2 143
BC003789.1 - 2 139 3593052
U17297.1 - 2 176
X91043.1 - 2 176

View Full Product Details